Learn More
Med Vet International Dynarex Ultrasound Gel, 8.5 Fl. oz, Clear, 12/Case
Catalog No. 502828961
Encompass_Preferred
Dynarex Ultrasound Gel is a premium electroconductive gel used in ultrasonic echo imaging systems. This Ultrasound Gel is acoustically correct for a wide range of frequencies. The non-greasy aqueous Gel is designed to not stain clothing or damage transducers. Ultrasound Gel is non-irritating and fragrance free for improved comfort. Bacteriostatic formula crafted to help prevent the growth of bacteria for added protection. Available in Blue or Clear and designed for easy refill.  Dynarex Ultrasound Gel is a non-greasy and hypoallergenic gel formulated to efficiently facilitate ultrasound procedures. This electroconductive Ultrasound Gel is acoustically correct for a broad range of frequencies.  
Catalog No. 50-282-8961 Supplier Med Vet International Supplier No. 1245
Dynarex Ultrasound Gel is a premium electroconductive gel used in ultrasonic echo imaging systems. This Ultrasound Gel is acoustically correct for a wide range of frequencies. The non-greasy aqueous Gel is designed to not stain clothing or damage transducers. Ultrasound Gel is non-irritating and fragrance free for improved comfort. Bacteriostatic formula crafted to help prevent the growth of bacteria for added protection. Available in Blue or Clear and designed for easy refill.  Dynarex Ultrasound Gel is a non-greasy and hypoallergenic gel formulated to efficiently facilitate ultrasound procedures. This electroconductive Ultrasound Gel is acoustically correct for a broad range of frequencies.  