Learn More
Med Vet International Ultrasound Gel Aquasonic Clear Transmission 8.5oz Bottle, 03-08, 12/Box
Catalog No. 502829216
Encompass_Preferred
No fragrance or color For diagnostic and therapeutic medical ultrasound procedures Acoustically correct for the broad range of frequencies used Hypoallergenic, bacteriostatic, nonirritating Water soluble, No formaldehyde, Not a spermicide No fragrance or color 5 liter SONICPAC with refillable dispenser
Catalog No. 50-282-9216 Supplier Med Vet International Supplier No. 227100BX
No fragrance or color For diagnostic and therapeutic medical ultrasound procedures Acoustically correct for the broad range of frequencies used Hypoallergenic, bacteriostatic, nonirritating Water soluble, No formaldehyde, Not a spermicide No fragrance or color 5 liter SONICPAC with refillable dispenser