Learn More
Med Vet International McKesson Ultrasound Gel, Clear, 5 Liters, Cubitainer, Ultrasound and Laser Transmission, 4964, Each
Catalog No. 503043795
Encompass_Preferred
Ultrasound Gel McKesson Transmission 5 Liter Jar This 5-liter McKesson ultrasound gel has a high viscosity to prevent running and to keep it in place after application The greaseless and water-soluble formula offers easy cleanup High conductivity delivers clearer images The non-corrosive high viscosity ultrasound gel is safe for equipment and instruments The gel is bacteriostatic to protect against cross contamination Free of fragrances, alcohol and salt, it's suitable for sensitive skin
Catalog No. 50-304-3795 Supplier Med Vet International Supplier No. 875484EA
Ultrasound Gel McKesson Transmission 5 Liter Jar This 5-liter McKesson ultrasound gel has a high viscosity to prevent running and to keep it in place after application The greaseless and water-soluble formula offers easy cleanup High conductivity delivers clearer images The non-corrosive high viscosity ultrasound gel is safe for equipment and instruments The gel is bacteriostatic to protect against cross contamination Free of fragrances, alcohol and salt, it's suitable for sensitive skin