Learn More
Med Vet International McKesson Ultrasound Gel, 5 Liter Cubitainer, Medium, NonSterile, 4962, 4/Case
Catalog No. 503045105
Encompass_Preferred
Ultrasound Gel McKesson Conductive Gel 5 Liter Jar McKesson Ultrasound Gel Pink 5 L (1.32 gal) Medium Viscosity Conductive, nonstaining, water soluble contact medium for ultrasound transmission Conductive feature for increased image clarity Water soluble for quick, easy clean-up Noncorrosive for extended instrument life Salt, alcohol, and fragrance free Bacteriostatic
Catalog No. 50-304-5105 Supplier Med Vet International Supplier No. 875464CS
Ultrasound Gel McKesson Conductive Gel 5 Liter Jar McKesson Ultrasound Gel Pink 5 L (1.32 gal) Medium Viscosity Conductive, nonstaining, water soluble contact medium for ultrasound transmission Conductive feature for increased image clarity Water soluble for quick, easy clean-up Noncorrosive for extended instrument life Salt, alcohol, and fragrance free Bacteriostatic