Learn More
Med Vet International McKesson Ultrasound Gel, 8.5 oz, 4961, Each
Catalog No. 503045163
Encompass_Preferred
Ultrasound Gel McKesson Conductive Gel 8.5 oz. Bottle Ultrasound gel to aide in coupling efficiency. This gel is water soluble for quick and easy cleanup after examinations8.5 oz (250 mL) Medium Viscosity Conductive, nonstaining, water soluble contact medium for ultrasound transmission Conductive feature for increased image clarity Water soluble for quick, easy clean-up noncorrosive for extended instrument life Salt, alcohol, and fragrance free Bacteriostatic
Catalog No. 50-304-5163 Supplier Med Vet International Supplier No. 875463EA
Ultrasound Gel McKesson Conductive Gel 8.5 oz. Bottle Ultrasound gel to aide in coupling efficiency. This gel is water soluble for quick and easy cleanup after examinations8.5 oz (250 mL) Medium Viscosity Conductive, nonstaining, water soluble contact medium for ultrasound transmission Conductive feature for increased image clarity Water soluble for quick, easy clean-up noncorrosive for extended instrument life Salt, alcohol, and fragrance free Bacteriostatic