Learn More
Med Vet International McKesson Ultrasound Gel, Clear, 250 mL (8.5 oz.) Bottle, Ultrasound and Laser Transmission, 4963, 12/Case
Catalog No. 503045084
Encompass_Preferred
Ultrasound Gel McKesson Transmission 8.5 oz. Bottle This 250-milliliter McKesson ultrasound gel has a high viscosity to prevent running and to keep it in place after application The greaseless and water-soluble formula offers easy cleanup High conductivity delivers clearer images The non-corrosive high viscosity ultrasound gel is safe for equipment and instruments The gel is bacteriostatic to protect against cross contamination Free of fragrances, alcohol and salt, it's suitable for sensitive skin
Catalog No. 50-304-5084 Supplier Med Vet International Supplier No. 875465CS
Ultrasound Gel McKesson Transmission 8.5 oz. Bottle This 250-milliliter McKesson ultrasound gel has a high viscosity to prevent running and to keep it in place after application The greaseless and water-soluble formula offers easy cleanup High conductivity delivers clearer images The non-corrosive high viscosity ultrasound gel is safe for equipment and instruments The gel is bacteriostatic to protect against cross contamination Free of fragrances, alcohol and salt, it's suitable for sensitive skin