Learn More
Med Vet International Dynarex L-Mesitran Soft Wound Gel, 1.75oz Tube, 24/Case
Catalog No. 502831067
Encompass_Preferred
Dynarex L-Mesitran Soft Wound Gel with Medical Grade Honey is a primary dressing gel designed to create a moist healing environment for a variety of wounds. L-Mesitran Soft is formulated with 40% medical-grade honey, hypo-allergenic lanolin, and vitamins C & E to retain hydration and protect skin at the wound site.  Formulated with 40% medical-grade honey, hypo-allergenic lanolin, and vitamins C and E to promote optimal healing of chronic wounds Moisturizing formula helps to restore wound pH balance Anti-microbial gel helps to cleanse and debride wounds to accelerate healing process Commonly used with secondary dressings to minimize adherence to the wound bed Ideal for use on chronic and acute wounds at stages 1, 2, 3, or 4
Catalog No. 50-283-1067 Supplier Med Vet International Supplier No. 3078
Dynarex L-Mesitran Soft Wound Gel with Medical Grade Honey is a primary dressing gel designed to create a moist healing environment for a variety of wounds. L-Mesitran Soft is formulated with 40% medical-grade honey, hypo-allergenic lanolin, and vitamins C & E to retain hydration and protect skin at the wound site.  Formulated with 40% medical-grade honey, hypo-allergenic lanolin, and vitamins C and E to promote optimal healing of chronic wounds Moisturizing formula helps to restore wound pH balance Anti-microbial gel helps to cleanse and debride wounds to accelerate healing process Commonly used with secondary dressings to minimize adherence to the wound bed Ideal for use on chronic and acute wounds at stages 1, 2, 3, or 4