Learn More
ILEX LIFE SCIENCES LLC GELACELL GELATIN 10X10CM SHEET
SDP
Supplier:  ILEX LIFE SCIENCES LLC GC0601RNSCC
Encompass_Preferred
Gelacell - Gelatin 10x10 cm nanofibrous scaffold sheet for 3D cell culture. INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications. DESCRIPTION: This product is a 10x10 cm scaffold sheet made from pharmaceutical grade gelatin that offers a 3D structure allowing cells to grow in a more physiologically relevant manner, promoting cell-cell interactions, tissue-like organization, and cell polarization. It can be cut into desired shape and sizes depending on experimental needs and setups. The scaffold is tailored for the cultivation of complex systems like cell microenvironments, tissue regeneration, and organoids. The optimized design of the scaffold ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thus preventing cellular waste build up.
Catalog No. 50-273-3945