Learn More
ILEX LIFE SCIENCES LLC Gelacell Haloscreening PLLA Scaffolds 96 well plate 1/EA
SDP
Catalog No. 5000580388
Encompass_Preferred
Gelacell HaloScreening - PLLA Scaffolds in 96-well Cell Culture Plate for HTS (1 plate) HaloScreening by Gelatex Technologies a ready-to-use 96-well cell culture plate embedded with microfibrous 3D scaffolds produced using patented HaloSpin technology The plate is specifically designed for high-throughput screening (HTS) in industrial or research settings The bottom of each well contains scaffolds made of high-quality poly-L-lactic acid (PLLA) with optimized fiber diameters for improved cell-cell and cell-matrix interactions The highly porous 3D structure of the scaffolds closely mimics the natural extracellular matrix Sterilized and ready to use the plate seamlessly integrates into standard workflows and automated systems Downstream analysis and visualization can be conducted directly in the plate used for cell culturing streamlining workflows and enhancing efficiency in HTS This integrated approach accelerates processes and improves productivity in industrial settings
Catalog No. 50-005-80388 Supplier ILEX LIFE SCIENCES LLC Supplier No. GC0805RNWP96A
Gelacell HaloScreening - PLLA Scaffolds in 96-well Cell Culture Plate for HTS (1 plate) HaloScreening by Gelatex Technologies a ready-to-use 96-well cell culture plate embedded with microfibrous 3D scaffolds produced using patented HaloSpin technology The plate is specifically designed for high-throughput screening (HTS) in industrial or research settings The bottom of each well contains scaffolds made of high-quality poly-L-lactic acid (PLLA) with optimized fiber diameters for improved cell-cell and cell-matrix interactions The highly porous 3D structure of the scaffolds closely mimics the natural extracellular matrix Sterilized and ready to use the plate seamlessly integrates into standard workflows and automated systems Downstream analysis and visualization can be conducted directly in the plate used for cell culturing streamlining workflows and enhancing efficiency in HTS This integrated approach accelerates processes and improves productivity in industrial settings