Learn More
ILEX LIFE SCIENCES LLC GELACELL PCL-PLGA INSERTS 12W
SDP
Catalog No. 502733933
Encompass_Preferred
Gelacell - PCL:PLGA scaffold inserts for 12-well plate 3D cell culture (12 pack). INTRO: Gelacell is a unique, 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications. Designed as a non-woven, highly porous scaffold, Gelacell offers exceptional biocompatibility and non-toxicity across a variety of cell types. DESCRIPTION: Specially designed 12-well plate scaffold “inserts” made from pharmaceutical grade polycaprolactone + poly lactic-co-glycolic acid (PCL:PLGA ratio is 70:30) offer cell culture conditions for short-to-medium durations. Perfectly tailored for the cultivation of complex systems like cell microenvironments, tissue regeneration, and organoids. The optimized design of the scaffold ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thereby preventing cellular waste build up.
Catalog No. 50-273-3933 Supplier ILEX LIFE SCIENCES LLC Supplier No. GC0801RNIN12A
Add to Cart
Add to Cart
Gelacell - PCL:PLGA scaffold inserts for 12-well plate 3D cell culture (12 pack). INTRO: Gelacell is a unique, 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications. Designed as a non-woven, highly porous scaffold, Gelacell offers exceptional biocompatibility and non-toxicity across a variety of cell types. DESCRIPTION: Specially designed 12-well plate scaffold “inserts” made from pharmaceutical grade polycaprolactone + poly lactic-co-glycolic acid (PCL:PLGA ratio is 70:30) offer cell culture conditions for short-to-medium durations. Perfectly tailored for the cultivation of complex systems like cell microenvironments, tissue regeneration, and organoids. The optimized design of the scaffold ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thereby preventing cellular waste build up.