Learn More
ILEX LIFE SCIENCES LLC GELACELL PCLPLGA 10X10CM SHEET
SDP
Catalog No. 502733931
Encompass_Preferred
Gelacell - PCL:PLGA 10x10 cm nanofibrous scaffold sheet for 3D cell culture. INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold designed for advanced in vitro 3D cell culture and tissue engineering applications. DESCRIPTION: This product is a 10x10 cm sheet made from pharmaceutical grade polycaprolactone + poly lactic-co-glycolic acid (PCL:PLGA ratio is 70:30) that offers a 3D structure allowing cells to grow in a more physiologically relevant manner, promoting cell-cell interactions, tissue-like organization, and cell polarization. It can be cut into desired shape and sizes depending on experimental needs. It is ideal for the cultivation of complex systems like cell microenvironments, tissue regeneration, and organoids. The optimized design ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thus preventing cellular waste build up.
Catalog No. 50-273-3931 Supplier ILEX LIFE SCIENCES LLC Supplier No. GC0801RNSCC
Gelacell - PCL:PLGA 10x10 cm nanofibrous scaffold sheet for 3D cell culture. INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold designed for advanced in vitro 3D cell culture and tissue engineering applications. DESCRIPTION: This product is a 10x10 cm sheet made from pharmaceutical grade polycaprolactone + poly lactic-co-glycolic acid (PCL:PLGA ratio is 70:30) that offers a 3D structure allowing cells to grow in a more physiologically relevant manner, promoting cell-cell interactions, tissue-like organization, and cell polarization. It can be cut into desired shape and sizes depending on experimental needs. It is ideal for the cultivation of complex systems like cell microenvironments, tissue regeneration, and organoids. The optimized design ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thus preventing cellular waste build up.