Learn More
ILEX LIFE SCIENCES LLC GELACELL PCLPLGA CELLCROWN 24W
SDP
Catalog No. 502733935
Encompass_Preferred
Gelacell - PCL:PLGA scaffolds fixed to cell crowns in 24-well plate for 3D cell culture (1 plate). INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications. DESCRIPTION: This variant of Gelacell is specially designed for medium-to-long term cell culture durations. Scaffolds made from pharmaceutical grade polycaprolactone + poly lactic-co-glycolic acid (PCL:PLGA ratio is 70:30) are fixed to cell crowns in a 24-well plate. The cell crowns act as Boyden chambers for facilitating studies related to air-liquid interface culture, co-culture, complex tissue models, etc. The optimized design ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thereby preventing cellular waste build up.
Catalog No. 50-273-3935 Supplier ILEX LIFE SCIENCES LLC Supplier No. GC0801RNCC24B
Add to Cart
Add to Cart
Gelacell - PCL:PLGA scaffolds fixed to cell crowns in 24-well plate for 3D cell culture (1 plate). INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications. DESCRIPTION: This variant of Gelacell is specially designed for medium-to-long term cell culture durations. Scaffolds made from pharmaceutical grade polycaprolactone + poly lactic-co-glycolic acid (PCL:PLGA ratio is 70:30) are fixed to cell crowns in a 24-well plate. The cell crowns act as Boyden chambers for facilitating studies related to air-liquid interface culture, co-culture, complex tissue models, etc. The optimized design ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thereby preventing cellular waste build up.