Learn More
ILEX LIFE SCIENCES LLC Gelacell PHB HYDRO Scaffold Inserts for 24 well plate 3D cell culture 1/EA
SDP
Catalog No. 5000580407
Encompass_Preferred
Gelacell - PHB HYDRO scaffold inserts for 24-well plate 3D cell culture (24 pack) INTRO Gelacell manufactured by Gelatex Technologies is a unique 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications Designed as a non-woven highly porous scaffold Gelacell offers exceptional biocompatibility and non-toxicity across a variety of cell types DESCRIPTION These hydrophilic 24-well plate scaffold inserts made from PHB HYDRO (blend of 91% polyhydroxybutyrate [PHB] and 9% polyethylene glycol [PEG]) can be fitted directly in 24 multi-well plates for short-to-medium cell culture durations Combining biocompatibility and robust mechanical stability with a sustainable bio-based composition Gelacell PHB HYDRO-Scaffold represents a novel platform for users seeking reliable scalable and eco-conscious solutions for next-generation 3D cell culture drug discovery and wound healing material development
Catalog No. 50-005-80407 Supplier ILEX LIFE SCIENCES LLC Supplier No. GC0901RNIN24A
Gelacell - PHB HYDRO scaffold inserts for 24-well plate 3D cell culture (24 pack) INTRO Gelacell manufactured by Gelatex Technologies is a unique 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications Designed as a non-woven highly porous scaffold Gelacell offers exceptional biocompatibility and non-toxicity across a variety of cell types DESCRIPTION These hydrophilic 24-well plate scaffold inserts made from PHB HYDRO (blend of 91% polyhydroxybutyrate [PHB] and 9% polyethylene glycol [PEG]) can be fitted directly in 24 multi-well plates for short-to-medium cell culture durations Combining biocompatibility and robust mechanical stability with a sustainable bio-based composition Gelacell PHB HYDRO-Scaffold represents a novel platform for users seeking reliable scalable and eco-conscious solutions for next-generation 3D cell culture drug discovery and wound healing material development