Learn More
ILEX LIFE SCIENCES LLC Gelacell PHB HYDRO Scaffold Sheet 10 x 10 cm 3D cell culture and tissue engineering 1/EA
SDP
Catalog No. 5000580375
Encompass_Preferred
Gelacell - PHB HYDRO 10x10 cm scaffold sheet for 3D cell culture INTRO Gelacell manufactured by Gelatex Technologies is a unique 3D microfibrous scaffold designed for advanced in vitro 3D cell culture and tissue engineering applications DESCRIPTION This hydrophilic 10 cm 10 cm scaffold sheet made from PHB HYDRO (blend of 91% polyhydroxybutyrate [PHB] and 9% polyethylene glycol [PEG]) can be used directly or customized for multiwell plates bioreactors or other experimental setups (it can be cut into desired shape and sizes) Combining biocompatibility mechanical stability and eco-friendly composition Gelacell PHB HYDRO provides researchers and industrial developers with a reliable scalable and next-generation platform for 3D cell culture drug discovery and wound healing material applications
Catalog No. 50-005-80375 Supplier ILEX LIFE SCIENCES LLC Supplier No. GC0901RNSCC
Gelacell - PHB HYDRO 10x10 cm scaffold sheet for 3D cell culture INTRO Gelacell manufactured by Gelatex Technologies is a unique 3D microfibrous scaffold designed for advanced in vitro 3D cell culture and tissue engineering applications DESCRIPTION This hydrophilic 10 cm 10 cm scaffold sheet made from PHB HYDRO (blend of 91% polyhydroxybutyrate [PHB] and 9% polyethylene glycol [PEG]) can be used directly or customized for multiwell plates bioreactors or other experimental setups (it can be cut into desired shape and sizes) Combining biocompatibility mechanical stability and eco-friendly composition Gelacell PHB HYDRO provides researchers and industrial developers with a reliable scalable and next-generation platform for 3D cell culture drug discovery and wound healing material applications