Learn More
ILEX LIFE SCIENCES LLC Gelacell PHB Scaffold Sheet 10 x 10 cm 3D cell culture and tissue engineering 1/EA
SDP
Catalog No. 5000580428
Encompass_Preferred
Gelacell - PHB 10x10 cm scaffold sheet for 3D cell culture INTRO Gelacell manufactured by Gelatex Technologies is a unique 3D microfibrous scaffold designed for advanced in vitro 3D cell culture and tissue engineering applications DESCRIPTION This 10 cm 10 cm scaffold sheet made from polyhydroxybutyrate (PHB) can be used directly or customized to fit a wide range of experimental setups including multiwell plates petri dishes and bioreactors (it can be cut into desired shapes and sizes) Combining biocompatibility and robust mechanical stability with a sustainable bio-based composition Gelacell PHB-Scaffold represents a novel platform for researchers and industrial developers seeking reliable scalable and eco-conscious solutions for next-generation 3D cell culture drug discovery and wound healing material development
Catalog No. 50-005-80428 Supplier ILEX LIFE SCIENCES LLC Supplier No. GC0900RNSCC
Gelacell - PHB 10x10 cm scaffold sheet for 3D cell culture INTRO Gelacell manufactured by Gelatex Technologies is a unique 3D microfibrous scaffold designed for advanced in vitro 3D cell culture and tissue engineering applications DESCRIPTION This 10 cm 10 cm scaffold sheet made from polyhydroxybutyrate (PHB) can be used directly or customized to fit a wide range of experimental setups including multiwell plates petri dishes and bioreactors (it can be cut into desired shapes and sizes) Combining biocompatibility and robust mechanical stability with a sustainable bio-based composition Gelacell PHB-Scaffold represents a novel platform for researchers and industrial developers seeking reliable scalable and eco-conscious solutions for next-generation 3D cell culture drug discovery and wound healing material development