Learn More
ILEX LIFE SCIENCES LLC GELACELL PLGA 10X10 CM SHEET
SDP
Supplier:  ILEX LIFE SCIENCES LLC GC0800RNSCC
Encompass_Preferred
Gelacell - PLGA 10x10 cm nanofibrous scaffold sheet for 3D cell culture. INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications. DESCRIPTION: This product is a 10x10 cm scaffold sheet made from pharmaceutical grade poly lactic-co-glycolic acid (PLGA) that offers a 3D structure allowing cells to grow in a more physiologically relevant manner, promoting cell-cell interactions, tissue-like organization, and cell polarization. It can be cut into desired shape/size depending on experimental needs and setups. The scaffold is tailored for cultivation of complex systems like cell microenvironments, tissue regeneration, and organoids. The optimized design ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thus preventing cellular waste build up.
Catalog No. 50-273-3938