Learn More
ILEX LIFE SCIENCES LLC GELACELL PLGA INSERTS 24WELL
SDP
Catalog No. 502733939
Encompass_Preferred
Gelacell - PLGA scaffold inserts for 24-well plate 3D cell culture (24 pack). INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications. Designed as a non-woven, highly porous scaffold, Gelacell offers exceptional biocompatibility and non-toxicity across a variety of cell types. DESCRIPTION: Specially designed 24-well plate scaffold “inserts” made from pharmaceutical grade poly lactic-co-glycolic acid (PLGA) offer cell culture conditions for short-to-medium durations. Perfectly tailored for the cultivation of complex systems like cell microenvironments, tissue regeneration, and organoids. The optimized design ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thereby preventing cellular waste build up.
Catalog No. 50-273-3939 Supplier ILEX LIFE SCIENCES LLC Supplier No. GC0800RNIN24A
Gelacell - PLGA scaffold inserts for 24-well plate 3D cell culture (24 pack). INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications. Designed as a non-woven, highly porous scaffold, Gelacell offers exceptional biocompatibility and non-toxicity across a variety of cell types. DESCRIPTION: Specially designed 24-well plate scaffold “inserts” made from pharmaceutical grade poly lactic-co-glycolic acid (PLGA) offer cell culture conditions for short-to-medium durations. Perfectly tailored for the cultivation of complex systems like cell microenvironments, tissue regeneration, and organoids. The optimized design ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thereby preventing cellular waste build up.