Learn More
ILEX LIFE SCIENCES LLC GELACELL PLLA 10X10CM SHEET
SDP
Supplier:  ILEX LIFE SCIENCES LLC GC0805RNSCC
Encompass_Preferred
Gelacell - PLLA 10x10 cm scaffold sheet for 3D cell culture. INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications. DESCRIPTION: This product is a 10x10 cm scaffold sheet made from pharmaceutical grade poly-L-lactide (PLLA) that offers a 3D structure allowing cells to grow in a more physiologically relevant manner, promoting cell-cell interactions, tissue-like organization, and cell polarization. It can be cut into desired shape/sizes depending on experimental needs. The scaffold is ideal for cultivation of complex systems like cell microenvironments, tissue regeneration, and organoids. The optimized design ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thereby preventing cellular waste build up.
Catalog No. 50-273-3918