Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotang Inc GELATIN (10%)., 125 ml

Catalog No. 50152200 Shop All BIOTANG Inc Products
Encompass_Preferred

GELATIN (10%)., 125 ml.

Catalog No. 50-152-200 Supplier Biotang Inc Supplier No. BTBB747
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GELATIN (10%)., 125 ml.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.