Learn More
Biotang Inc GELATIN 10% 125ML. IBB-184. 10% Gelatin. Prepared in 18.2 megohms water. Shipped on blue ice. Store at -20 degree Celsius.
Catalog No. 507513804 Shop All BIOTANG Inc Products
Encompass_Preferred
GELATIN 10% 125ML. IBB-184. 10% Gelatin. Prepared in 18.2 megohms water. Shipped on blue ice. Store at -20 degree Celsius.
Catalog No. 50-751-3804 Supplier Biotang Inc Supplier No. BTBB105
Add to Cart
Add to Cart
GELATIN 10% 125ML. IBB-184. 10% Gelatin. Prepared in 18.2 megohms water. Shipped on blue ice. Store at -20 degree Celsius.