Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gelatin, 10% soln., Thermo Scientific Chemicals

Catalog No. AAJ62699AK
Change view
Click to view available options
Quantity:
125 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62699AK 250 mL
AAJ62699EQE 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62699AK Supplier Thermo Scientific Chemicals Supplier No. J62699AK
  • $281.00
    $215.65 / Each of 1

    Save $65.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

Gelatin is used for preparing slides for histochemistry and light microscope autoradiography. Gelatin has also been added to cell culture plates and improves the attachment of a variety of cell types. Additionally, gelatin can inhibit crystallization in bacteriology studies. In addition it is an excellent diluents for primary and secondary antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-70-8
MDL Number MFCD00081638

Specifications

Concentration 10% soln.
Solubility Information Soluble in water (50mg/ml),glycerol,and acetic acid.
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material Gelatin
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.