Learn More
Alkali Scientific Gelatin 10% Solution, 100 Milliliters
SDP
Catalog No. 5000659737
Encompass_Preferred
Gelatin 10% Solution, 100 Milliliters, is a laboratory reagent widely used as a stabilizer and gelling agent in various biological and chemical applications. This solution is ideal for use in microbiological media, electrophoresis, and in pharmaceutical formulations. Gelatin is commonly used to create culture media for growing bacteria, fungi, and cells, as it provides a solid surface for microbial growth while maintaining the necessary moisture content. It is also used in protein research and as a stabilizer for enzymes and vaccines. The 10% concentration offers an optimal balance for laboratory use, ensuring effective gelling without excessive viscosity.
Catalog No. 50-006-59737 Supplier Alkali Scientific Supplier No. LAT001CP
Gelatin 10% Solution, 100 Milliliters, is a laboratory reagent widely used as a stabilizer and gelling agent in various biological and chemical applications. This solution is ideal for use in microbiological media, electrophoresis, and in pharmaceutical formulations. Gelatin is commonly used to create culture media for growing bacteria, fungi, and cells, as it provides a solid surface for microbial growth while maintaining the necessary moisture content. It is also used in protein research and as a stabilizer for enzymes and vaccines. The 10% concentration offers an optimal balance for laboratory use, ensuring effective gelling without excessive viscosity.