Learn More
Research Products International Corp Gelatin, 100 Grams

Supplier: Research Products International Corp G05100100.0
- CAS Number: 9000-70-8
- Soluble in: Water
- Store at: Room Temperature
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.