Learn More
Crescent Chemical Co Inc GELATIN
SDP
Supplier:  Crescent Chemical Co Inc SE22151.02
Encompass_Preferred
Gelatin Special quality for bacteriology. A 5 % solution liquefies at 25 - 30 °C and begins to set at 19 - 25 °C. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 22151.02 CAS 9000-70-8
Catalog No. 50-247-614