Learn More
Alkali Scientific Gelatin, 500 Grams
SDP
Catalog No. 5000659802
Encompass_Preferred
Gelatin, 500 Grams, is a laboratory-grade biochemical reagent widely used as a protein source in microbiology, cell culture, and biochemistry. Gelatin is a natural protein derived from animal collagen and is used in media preparation to support the growth of microorganisms. In addition to its use in bacterial growth media, gelatin is also employed in electrophoresis, enzyme assays, and various other biochemical tests. The laboratory grade ensures that the gelatin meets the highest purity standards, making it suitable for sensitive and reproducible experiments. This 500g package is perfect for laboratories requiring a larger supply for ongoing research.
Catalog No. 50-006-59802 Supplier Alkali Scientific Supplier No. ELA001CP
Gelatin, 500 Grams, is a laboratory-grade biochemical reagent widely used as a protein source in microbiology, cell culture, and biochemistry. Gelatin is a natural protein derived from animal collagen and is used in media preparation to support the growth of microorganisms. In addition to its use in bacterial growth media, gelatin is also employed in electrophoresis, enzyme assays, and various other biochemical tests. The laboratory grade ensures that the gelatin meets the highest purity standards, making it suitable for sensitive and reproducible experiments. This 500g package is perfect for laboratories requiring a larger supply for ongoing research.