Learn More
Electron Microscopy Sciences Gelatin Capsule Holder for Size "00"
SDP
Catalog No. 5026082
Encompass_Preferred
Made from durable polyethylene. It accommodates 15 gelatin capsules size "00". The whole capsule is exposed to allow for even heat and/or UV light distribution during polymerization.
Catalog No. 50-260-82 Supplier Electron Microscopy Sciences Supplier No. 7012004
Made from durable polyethylene. It accommodates 15 gelatin capsules size "00". The whole capsule is exposed to allow for even heat and/or UV light distribution during polymerization.