Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Gelatin Capsule Size 0, Volume 0.68ml
SDP

Catalog No. 5024872
Encompass_Preferred

Natural, hard gelatin capsules. To be used as an embedding mold for water miscible resins, or resins which need to be cured by transmitted light. Gelatin capsules dissolve or disintegrate within 5 minutes when immersed in a 0.5% HCl solution at 36-38 degree C, within 2 minutes in water at 37 degree C or in artificial gastric juice.
OAL: 21.20 mm
Diameter: 7.34 mm
Volume: 0.68 mL

Catalog No. 50-248-72 Supplier Electron Microscopy Sciences Supplier No. 70110
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Natural, hard gelatin capsules. To be used as an embedding mold for water miscible resins, or resins which need to be cured by transmitted light. Gelatin capsules dissolve or disintegrate within 5 minutes when immersed in a 0.5% HCl solution at 36-38 degree C, within 2 minutes in water at 37 degree C or in artificial gastric juice.
OAL: 21.20 mm
Diameter: 7.34 mm
Volume: 0.68 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.