Learn More
Electron Microscopy Sciences Gelatin Capsule Size 12
SDP
Catalog No. 5026878
Encompass_Preferred
Our large size gelatin capsules are ideal for storing either delicate or small items. It is also suitable for use as an embedding mold for paraffin or paraplast. The capsules come complete with a locking ring.
Catalog No. 50-268-78 Supplier Electron Microscopy Sciences Supplier No. 70115
Our large size gelatin capsules are ideal for storing either delicate or small items. It is also suitable for use as an embedding mold for paraffin or paraplast. The capsules come complete with a locking ring.