Learn More
Electron Microscopy Sciences Gelatin Capsule Size 4
SDP
Catalog No. 5024873
Encompass_Preferred
Natural, hard gelatin capsules. To be used as an embedding mold for water miscible resins, or resins which need to be cured by transmitted light. Gelatin capsules dissolve or disintegrate within 5 minutes when immersed in a 0.5% HCl solution at 36-38 degree C, within 2 minutes in water at 37 degree C or in artificial gastric juice.
OAL: 13.90 mm
Diameter: 5.05 mm
Volume: 0.21 mL
Catalog No. 50-248-73 Supplier Electron Microscopy Sciences Supplier No. 70105
Natural, hard gelatin capsules. To be used as an embedding mold for water miscible resins, or resins which need to be cured by transmitted light. Gelatin capsules dissolve or disintegrate within 5 minutes when immersed in a 0.5% HCl solution at 36-38 degree C, within 2 minutes in water at 37 degree C or in artificial gastric juice.
OAL: 13.90 mm
Diameter: 5.05 mm
Volume: 0.21 mL