Learn More
Fd Neurotechnologies Inc GELATIN-COATED MICROSCOPE
SDP
Catalog No. NC0785009
Encompass
Highest quality glass and precleaned microscope slides are coated with 3% gelatin. Each slide (size: 75x25 mm, thickness: 1.0 mm) has a frosted write-on end. The slides are packaged in a premium quality slide box sealed in an air-tight plastic bag to ensure dryness during shipment and storage. 100 slides per box.
Catalog No. NC0785009 Supplier Fd Neurotechnologies Inc Supplier No. PO101
Highest quality glass and precleaned microscope slides are coated with 3% gelatin. Each slide (size: 75x25 mm, thickness: 1.0 mm) has a frosted write-on end. The slides are packaged in a premium quality slide box sealed in an air-tight plastic bag to ensure dryness during shipment and storage. 100 slides per box.