Learn More
Bio Rad Laboratories GELATIN EIA GRADE 200G
Catalog No. NC9294874
Encompass
Gelatin, EIA grade, 200g, Detergent/ Blocking Reagent to complete western blot experiments
Catalog No. NC9294874 Supplier Bio Rad Laboratories Supplier No. 1706537
Gelatin, EIA grade, 200g, Detergent/ Blocking Reagent to complete western blot experiments