Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gelatin, ( FCC), MP Biomedicals

Catalog No. ICN96031791 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN96031791 1 kg
ICN96031790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN96031791 Supplier MP Biomedicals, Inc Supplier No. 0296031791
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9000-70-8
MDL Number MFCD00081638

Specifications

Quantity 1 kg
Grade FCC
Chemical Name or Material Gelatin, Type A; 100 Bloom
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.