Learn More
Bioworld Gelatin for Bacteriological Media, 500 g
Supplier:  Bioworld 407000161
Encompass_Preferred
A high grade granular preparation of gelatin that can be used alone as a solidification agent or incorporated into microbiological media as a nutrient or to determine gelatin liquefaction. Specifications:Ash: 0.17%Moisture: 8.7%Coliforms: less than 3/gmStd. Plate: 10/gmE. coli: None DetectedSalmonella: None DetectedAppearance: SolidBoiling Point : 100 °CDensity : 0.68 g/cm3 at 20 °CHS Code : 35030010PH Value: 5.0RTECS Number: LX8580000Viscosity: 46 MPS
Catalog No. 50-256-022