Learn More
Crescent Chemical Co Inc Gelatin, liquid, MFCD00081638, 100mL
SDP
Supplier:  Crescent Chemical Co Inc SE22156.01
Encompass_Preferred
From fish skin. Solids content 44.0 - 46.0 %. Contains methyl-/propyl-PHB as preservative. Specially prepared and purified gelatin. Supplied as a pourable liquid. Completely water soluble, even at room temperature and at high concentrations. Acts as a protective colloid. Storage Conditions: +15 °C TO +30 °C.
Catalog No. 50-247-615