Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gelatin Powder, Lab Grade, Innovating Science™
SCI_ED

Catalog No. S031112 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S031112 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S031112 Supplier Aldon Corporation Supplier No. IS16003
Add to Cart
Add to Cart

Specifications

Form Powder
Grade Laboratory
Manufacturing Origin United States
Product Type Gelatin
Sterility Non-sterile
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.