Learn More
ILEX LIFE SCIENCES LLC GELATIN SOLUTION 0.1 500ML
SDP
Catalog No. 502733729
Encompass_Preferred
PAN-Biotech Gelatin Solution 0.1% in DPBS, w/o: Ca, Mg (500 ml). DESCRIPTION: PAN-Biotech Gelatin Solution 0.1% in DPBS, w/o: Ca, Mg for laboratory research applications. Gelatin is a heterogenous mixture of water soluble proteins of high average molecular weight present in collagen. The proteins are extracted by boiling skin, tendons, ligaments, or bones in water. APPLICATION: Applications using gelatin include coating cell culture plates to improve cell attachment for a variety of cell types, addition to PCR to stabilize Taq DNA Polymerase, and use as a blocking agent in Western Blotting, ELISA, and immunohistochemistry. In bacteriology, gelatin can be used as a component of culture media. STORAGE: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany.
Catalog No. 50-273-3729 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0620410
PAN-Biotech Gelatin Solution 0.1% in DPBS, w/o: Ca, Mg (500 ml). DESCRIPTION: PAN-Biotech Gelatin Solution 0.1% in DPBS, w/o: Ca, Mg for laboratory research applications. Gelatin is a heterogenous mixture of water soluble proteins of high average molecular weight present in collagen. The proteins are extracted by boiling skin, tendons, ligaments, or bones in water. APPLICATION: Applications using gelatin include coating cell culture plates to improve cell attachment for a variety of cell types, addition to PCR to stabilize Taq DNA Polymerase, and use as a blocking agent in Western Blotting, ELISA, and immunohistochemistry. In bacteriology, gelatin can be used as a component of culture media. STORAGE: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany.