Learn More
ILEX LIFE SCIENCES LLC GELATIN SOLUTION 2.0 6 X 100ML
SDP
Catalog No. 502733726
Encompass_Preferred
PAN-Biotech Gelatin Solution 2% in DPBS, w/o: Ca, Mg (6 x 100 ml). DESCRIPTION: PAN-Biotech Gelatin Solution 2% in DPBS, w/o: Ca, Mg for laboratory research applications. Gelatin is a heterogenous mixture of water soluble proteins of high average molecular weight present in collagen. The proteins are extracted by boiling skin, tendons, ligaments, or bones in water. APPLICATION: Applications using gelatin include coating cell culture plates to improve cell attachment for a variety of cell types, addition to PCR to stabilize Taq DNA Polymerase, and use as a blocking agent in Western Blotting, ELISA, and immunohistochemistry. In bacteriology, gelatin can be used as a component of culture media. STORAGE: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany.
Catalog No. 50-273-3726 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0625200X6
Add to Cart
Add to Cart
PAN-Biotech Gelatin Solution 2% in DPBS, w/o: Ca, Mg (6 x 100 ml). DESCRIPTION: PAN-Biotech Gelatin Solution 2% in DPBS, w/o: Ca, Mg for laboratory research applications. Gelatin is a heterogenous mixture of water soluble proteins of high average molecular weight present in collagen. The proteins are extracted by boiling skin, tendons, ligaments, or bones in water. APPLICATION: Applications using gelatin include coating cell culture plates to improve cell attachment for a variety of cell types, addition to PCR to stabilize Taq DNA Polymerase, and use as a blocking agent in Western Blotting, ELISA, and immunohistochemistry. In bacteriology, gelatin can be used as a component of culture media. STORAGE: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany.