Learn More
Eurofins Discoverx Geldanamycin, 1 mg. (92-1235)
Supplier:  Eurofins Discoverx 921235
Encompass
Geldanamycin is an antibiotic and a potent inhibitor of the heat shock protein 90 (Hsp90) protein (KD=1.2uM), binding its ATP site and inhibiting its chaperone activity. Induces selective degradation of mutated versions of v-Src, Bcr-Abl, Raf, p53, and steroid receptors in tumor cells via Hsp90 inhibition. Affects multiple cellular processes, including gene expression, cell proliferation, apoptosis, and angiogenesis. Provided as 1 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9757