Learn More
Medchemexpress LLC HY-15230 10mg Medchemexpress, Geldanamycin CAS:30562-34-6 Purity:>98%
SDP
Catalog No. 502029103
Encompass_Preferred
Medchemexpress, HY-15230 10mg Geldanamycin CAS:30562-34-6 Geldanamycin is a Hsp90 inhibitor with antimicrobial activity against many Gram-positive and some Gram-negative bacteria. Geldanamycin has anti-influenza virus H5N1 activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9103 Supplier Medchemexpress LLC Supplier No. HY1523010MG
Medchemexpress, HY-15230 10mg Geldanamycin CAS:30562-34-6 Geldanamycin is a Hsp90 inhibitor with antimicrobial activity against many Gram-positive and some Gram-negative bacteria. Geldanamycin has anti-influenza virus H5N1 activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.