Learn More
Apexbio Technology LLC Geldanamycin 30562-34-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064052
Encompass_Preferred
Geldanamycin is an antibiotic from Streptomyces hygroscopicus that inhibits heat shock protein 90 (Hsp90) by binding to its ATP-binding pocket This disruption affects protein folding and stabilization impacting cell proliferation stress response and tumor biology Geldanamycin is used as a molecular tool in these studies and has activity against various microbes including bacteria fungi protozoa and nematodes IC50 values range from nanomolar to micromolar levels depending on the cell line and conditions It is also applied in research on vascular relaxation glucocorticoid receptor modulation and inflammation-related signaling pathways
Catalog No. 50-000-64052 Supplier Apexbio Technology LLC Supplier No. A40605.1
Geldanamycin is an antibiotic from Streptomyces hygroscopicus that inhibits heat shock protein 90 (Hsp90) by binding to its ATP-binding pocket This disruption affects protein folding and stabilization impacting cell proliferation stress response and tumor biology Geldanamycin is used as a molecular tool in these studies and has activity against various microbes including bacteria fungi protozoa and nematodes IC50 values range from nanomolar to micromolar levels depending on the cell line and conditions It is also applied in research on vascular relaxation glucocorticoid receptor modulation and inflammation-related signaling pathways