Learn More
Medchemexpress LLC Geldanamycin | 30562-34-6 | C29H40N2O9 | 100 MG
SDP
Catalog No. 5000373107
Encompass_Preferred
Geldanamycin is an analytical standard intended for research and analytical applications. It acts as an Hsp90 inhibitor, demonstrating antimicrobial activity against various Gram-positive and some Gram-negative bacteria. Additionally, Geldanamycin has shown efficacy in inhibiting the H5N1 influenza virus.
  • Inhibits Hsp90
  • Possesses antimicrobial activity against Gram-positive and some Gram-negative bacteria
  • Exhibits anti-influenza virus H5N1 activities
Catalog No. 50-003-73107 Supplier Medchemexpress LLC Supplier No. HY15230100MG
Geldanamycin is an analytical standard intended for research and analytical applications. It acts as an Hsp90 inhibitor, demonstrating antimicrobial activity against various Gram-positive and some Gram-negative bacteria. Additionally, Geldanamycin has shown efficacy in inhibiting the H5N1 influenza virus.
  • Inhibits Hsp90
  • Possesses antimicrobial activity against Gram-positive and some Gram-negative bacteria
  • Exhibits anti-influenza virus H5N1 activities