Learn More
Medchemexpress LLC Geldanamycin | 30562-34-6 | C29H40N2O9 | 50 MG
SDP
Catalog No. 5000373253
Encompass_Preferred
Geldanamycin is a potent Hsp90 inhibitor derived from *Streptomyces hygroscopicus*, exhibiting significant antimicrobial activity against various bacteria and anti-influenza virus H5N1 properties. It acts by binding to the N-terminal ATPase domain of Hsp90, thereby inhibiting its chaperone function and inducing apoptotic tumor cell death.
  • Hsp90 inhibitor
  • Antimicrobial activity
  • Anti-influenza virus H5N1 activities
  • Induces tumor cell death
  • Light yellow to orange solid appearance
  • High purity (99.43%)
Catalog No. 50-003-73253 Supplier Medchemexpress LLC Supplier No. HY1523050MG
Geldanamycin is a potent Hsp90 inhibitor derived from *Streptomyces hygroscopicus*, exhibiting significant antimicrobial activity against various bacteria and anti-influenza virus H5N1 properties. It acts by binding to the N-terminal ATPase domain of Hsp90, thereby inhibiting its chaperone function and inducing apoptotic tumor cell death.
  • Hsp90 inhibitor
  • Antimicrobial activity
  • Anti-influenza virus H5N1 activities
  • Induces tumor cell death
  • Light yellow to orange solid appearance
  • High purity (99.43%)