Learn More
Selleck Chemical LLC Geldanamycin S2713-1g
SDP
Catalog No. 503112554
Encompass_Preferred
Geldanamycin is a natural existing HSP90 inhibitor with Kd of 1 2 M specifically disrupts glucocorticoid receptor (GR)/HSP association Geldanamycin attenuates virus infection-induced ALI (acute lung injury)/ARDS (acute respiratory distress syndrome) by reducing the host s inflammatory responses
Catalog No. 50-311-2554 Supplier Selleck Chemical LLC Supplier No. S27131G
Geldanamycin is a natural existing HSP90 inhibitor with Kd of 1 2 M specifically disrupts glucocorticoid receptor (GR)/HSP association Geldanamycin attenuates virus infection-induced ALI (acute lung injury)/ARDS (acute respiratory distress syndrome) by reducing the host s inflammatory responses