Learn More
TARGETMOL CHEMICALS INC GELDANAMYCIN 50MG
SDP
Catalog No. 502576527
Encompass_Preferred
Also available in 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Geldanamycin an HSP90 inhibitor (Kd 1.2 uM) specifically disrupts glucocorticoid receptor (GR)/HSP association. purity: 98%
Catalog No. 50-257-6527 Supplier TARGETMOL CHEMICALS INC Supplier No. T634350MG
Also available in 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Geldanamycin an HSP90 inhibitor (Kd 1.2 uM) specifically disrupts glucocorticoid receptor (GR)/HSP association. purity: 98%