Learn More
Enzo Life Sciences Geldanamycin-biotin (1mg)
SDP
Supplier:  Enzo Life Sciences BMLEI3410001
Encompass_Preferred
A useful tool for affinity purification of HSP-90 and HSP-90 dependent client proteins. Purity: ≥98%. Long Term Storage: -20°C.
Catalog No. 50-201-0725