Learn More
Enzo Life Sciences Geldanamycin-FITC (1mg)
SDP
Supplier:  Enzo Life Sciences BMLEI3610001
Encompass_Preferred
A novel geldanamycin fluorescent probe that may be used in a fluorescence polarization assay for HSP90 inhibitors. May also be used for detection of cell surface HSP90. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0744