Learn More
Medchemexpress LLC Geldanamycin-FITC | 2969156-01-0 | 1034.18 | 1 MG
SDP
Catalog No. 5000365120
Encompass_Preferred
Geldanamycin-FITC is a fluorescent probe derived from Geldanamycin. It is utilized in fluorescence polarization assays for HSP90 inhibitors and can also detect cell surface HSP90.
  • Fluorescent probe
  • Used in fluorescence polarization assays for HSP90 inhibitors
  • Detects cell surface HSP90
  • Excitation (Ex): 488 nm
  • Emission (Em): 515 nm
Catalog No. 50-003-65120 Supplier Medchemexpress LLC Supplier No. 1MGHY133705
Geldanamycin-FITC is a fluorescent probe derived from Geldanamycin. It is utilized in fluorescence polarization assays for HSP90 inhibitors and can also detect cell surface HSP90.
  • Fluorescent probe
  • Used in fluorescence polarization assays for HSP90 inhibitors
  • Detects cell surface HSP90
  • Excitation (Ex): 488 nm
  • Emission (Em): 515 nm