Learn More
Medchemexpress LLC Geldanamycin-FITC | 2969156-01-0 | MFCD32068377 | 98.4% | C55H63N5O13S | 10MG
SDP
Catalog No. 5000258339
Encompass_Preferred
Geldanamycin-FITC is a fluorescein (FITC)-labeled derivative of geldanamycin used as a fluorescent probe for HSP90. It is suitable for fluorescence polarization assays and for detection of cell-surface HSP90, with excitation/emission at 488/515 nm.
  • Fluorescein (FITC)-labeled geldanamycin probe.
  • Designed for fluorescence polarization assays.
  • Detects cell-surface HSP90 (Ex/Em = 488/515 nm).
  • High purity (~98.4%).
  • Molecular formula C55H63N5O13S; molecular weight 1,034.18.
  • Available in multiple small-molecule pack sizes.
Catalog No. 50-002-58339 Supplier Medchemexpress LLC Supplier No. HY13370510MG
Geldanamycin-FITC is a fluorescein (FITC)-labeled derivative of geldanamycin used as a fluorescent probe for HSP90. It is suitable for fluorescence polarization assays and for detection of cell-surface HSP90, with excitation/emission at 488/515 nm.
  • Fluorescein (FITC)-labeled geldanamycin probe.
  • Designed for fluorescence polarization assays.
  • Detects cell-surface HSP90 (Ex/Em = 488/515 nm).
  • High purity (~98.4%).
  • Molecular formula C55H63N5O13S; molecular weight 1,034.18.
  • Available in multiple small-molecule pack sizes.