Learn More
Abcam Geldanamycin, Hsp90 inhibitor, 5MG
Catalog No. 502707154
Encompass_Preferred
MW 560.64 g/mol, Purity >98%. Selective Hsp90 inhibitor. Binds the ATP site of Hsp90 and inhibits chaperone activity. A benzoquinone ansamycin antibiotic. Also inhibits the Hsp90 paralog GRP94. Exhibits various cellular actions including gene expression, cell proliferation, apoptosis and angiogenesis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7154 Supplier Abcam Supplier No. AB1208965MG
MW 560.64 g/mol, Purity >98%. Selective Hsp90 inhibitor. Binds the ATP site of Hsp90 and inhibits chaperone activity. A benzoquinone ansamycin antibiotic. Also inhibits the Hsp90 paralog GRP94. Exhibits various cellular actions including gene expression, cell proliferation, apoptosis and angiogenesis.
The product is subject to the following: Abcam Restricted Use Statement