Learn More
Alkali Scientific Geldanamycin, from Streptomyces hygroscopicus, 1 Mg
SDP
Catalog No. 5000660230
Encompass_Preferred
Geldanamycin, from Streptomyces hygroscopicus, is a well-known inhibitor of heat shock protein 90 (Hsp90), a key molecular chaperone involved in the stabilization of several client proteins. This 1 mg formulation is widely used in protein research, particularly in studies related to cancer and neurodegenerative diseases. By inhibiting Hsp90, Geldanamycin accelerates the degradation of client proteins, including those involved in oncogenic signaling. It is also being studied as a potential therapeutic agent in targeted cancer treatments. This compound is critical in research investigating the role of Hsp90 in protein folding, cell signaling, and disease progression.
Catalog No. 50-006-60230 Supplier Alkali Scientific Supplier No. GEL002CP
Geldanamycin, from Streptomyces hygroscopicus, is a well-known inhibitor of heat shock protein 90 (Hsp90), a key molecular chaperone involved in the stabilization of several client proteins. This 1 mg formulation is widely used in protein research, particularly in studies related to cancer and neurodegenerative diseases. By inhibiting Hsp90, Geldanamycin accelerates the degradation of client proteins, including those involved in oncogenic signaling. It is also being studied as a potential therapeutic agent in targeted cancer treatments. This compound is critical in research investigating the role of Hsp90 in protein folding, cell signaling, and disease progression.